Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLUAP1 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CLUAP1 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1591
![]() ![]() |
Proteintech Group Inc
174701AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
CLUAP1 may play a role in cell proliferation or apoptosis.Specifications
CLUAP1 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Canine, Human, Mouse, Rat | |
Q6AYJ5, Q8R3P7, Q96AJ1 | |
23059, 363544, 479868, 76779 | |
CLUAP1 Fusion Protein Ag11419 | |
Primary | |
-20°C | |
CLUAP1 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CLUAP1, clusterin associated protein 1, KIAA0643, Qilin | |
Cluap1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title