Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLUH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571324
Description
CLUH Polyclonal Antibody for Western Blot
mRNA-binding protein involved in proper cytoplasmic distribution of mitochondria. Specifically binds mRNAs of nuclear-encoded mitochondrial proteins in the cytoplasm and regulates transport or translation of these transcripts close to mitochondria, playing a role in mitochondrial biogenesis.
Specifications
CLUH | |
Polyclonal | |
Unconjugated | |
CLUH | |
1300001I01Rik; cb498; CG8443-like; CLU1; cluh; cluha; clustered mitochondria (cluA/CLU1) homolog; clustered mitochondria (cluA/CLU1) homolog a; clustered mitochondria homolog; clustered mitochondria protein homolog; KIAA0664; LOW QUALITY PROTEIN: clustered mitochondria protein homolog; mKIAA0664; RGD1307222; sb:cb498; Unknown (protein for IMAGE:6905374); wu:fb77e08; wu:fd36f07; wu:fe05g06; wu:fi27b10; zgc:152873 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23277 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75153 | |
CLUH | |
synthetic peptide directed towards the C-terminal region of human CLUH. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction