Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLUH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109559 Supplier Invitrogen™ Supplier No. PA5109559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDLSLRFLEN ALAVSTKYHG PKALKVALSH HLVARVYESK AEFRSALQHE KEGYTIYKTQ LGEDHEKTKE SSEYLKCLTQ QAVALQRTMN EIYRNGSSAN IPPLKFTAPS MASVLEQLNV INGILFIPLS QKDLENLKAE VARRHQLQEA SRNRDRAEEP MATEPAPAGA PGDLGSQPPA AKDPSPSVQG.

mRNA-binding protein involved in proper cytoplasmic distribution of mitochondria. Specifically binds mRNAs of nuclear-encoded mitochondrial proteins in the cytoplasm and regulates transport or translation of these transcripts close to mitochondria, playing a role in mitochondrial biogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLUH
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLUH
Gene Accession No. O75153, Q5SW19
Gene Alias 1300001I01Rik; cb498; CG8443-like; CLU1; cluh; cluha; clustered mitochondria (cluA/CLU1) homolog; clustered mitochondria (cluA/CLU1) homolog a; clustered mitochondria homolog; clustered mitochondria protein homolog; KIAA0664; LOW QUALITY PROTEIN: clustered mitochondria protein homolog; mKIAA0664; RGD1307222; sb:cb498; Unknown (protein for IMAGE:6905374); wu:fb77e08; wu:fd36f07; wu:fe05g06; wu:fi27b10; zgc:152873
Gene Symbols CLUH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1120-1309 of human CLUH (NP_056044.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23277, 74148
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.