Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clusterin/APOJ Antibody (Hs-3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500552H
Description
Clusterin/APOJ Monoclonal antibody specifically detects Clusterin/APOJ in Human, Bovine (Negative), Canine (Negative), Feline (Negative), Porcine (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Clusterin/APOJ | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Bovine (Negative), Canine (Negative), Feline (Negative), Porcine (Negative) | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Hs-3 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2 | |
Freshly ejaculated human sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid and subsequently used for immunization. | |
0.1 mL | |
Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity | |
1191 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction