Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clusterin/APOJ Antibody (SAIC-43B-8), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320109F
Description
Clusterin/APOJ Monoclonal antibody specifically detects Clusterin/APOJ in Human samples. It is validated for ELISASpecifications
Clusterin/APOJ | |
Monoclonal | |
FITC | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2 | |
Peptide "e;ASSIIDELFQDR"e; derived from Clusterin conjugated to KLH. Clusterin isoform 1 functions as extracellular chaperone that prevents aggregation of nonnative proteins | |
0.1 mL | |
Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity | |
1191 | |
Store at 4°C in the dark. | |
IgG κ |
ELISA | |
SAIC-43B-8 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction