Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clusterin/APOJ Rabbit anti-Human, Clone: 210, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $548.00
Specifications
Antigen | Clusterin/APOJ |
---|---|
Clone | 210 |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Clusterin/APOJ Monoclonal specifically detects Clusterin/APOJ in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Clusterin/APOJ | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2 | |
Recombinant Human Clusterin/APOJ protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
210 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity | |
0.2 um filtered solution in PBS | |
1191 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title