Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLVS2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$173.10 - $403.50
Specifications
Antigen | CLVS2 |
---|---|
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CLVS2 Polyclonal antibody specifically detects CLVS2 in Human samples. It is validated for Western BlotSpecifications
CLVS2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS with 50% glycerol, pH7.3. | |
134829 | |
IgG | |
Affinity purified |
Western Blot 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
bA160A10.4;C6orf212;C6orf213;Clavesin-2;RLBP1L2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 265-327 of human CLVS2 (NP_001010852.2). LLDHEYDDDSEYNVDSYSMPVKEVEKELSPKSMKRSQSVVDPTVLKRMDKNEEENMQPLLSLD | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title