Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLVS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109560 Supplier Invitrogen™ Supplier No. PA5109560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLDHEYDDDS EYNVDSYSMP VKEVEKELSP KSMKRSQSVV DPTVLKRMDK NEEENMQPLL SLD.

Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLVS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLVS2
Gene Accession No. Q5SYC1
Gene Alias A330019N05Rik; bA160A10.4; C6orf212; C6orf213; clathrin vesicle-associated Sec14 protein 2; clavesin 2; clavesin-2; clavesin-2 {ECO:0000303; CLVS2; clvs2 {ECO:0000303; PubMed:19651769}; retinaldehyde binding protein 1-like 2; retinaldehyde-binding protein 1-like 2; Retinaldehyde-binding protein 1-like protein 2; RGD1306801; Rlbp1l2; si:ch211-199i15.4
Gene Symbols CLVS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 265-327 of human CLVS2 (NP_001010852.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134829
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.