Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ CM 10

Catalog No. 693310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-331-0 10 mg
69-335-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-331-0 Supplier Tocris™ Supplier No. 6933/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

ALDH1A inhibitor; depletes CD133+ cancer stem cells (CSC)

CM 10 is a aldehyde dehydrogenase 1A (ALDH1A) inhibitor (IC50 values are 0.64, 0.74 and 1.7 μM for ALDH1A3, ALDH1A2 and ALDH1A1, respectively). Induces a necroptotic phenotype and preferentially depletes CD133+ ovarian cancer stem cells in vitro.

Specifications

Quantity 10 mg
Molecular Formula C20H23N3O
Formula Weight 321.42
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.