Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589409 Supplier Invitrogen™ Supplier No. PA589409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDSVEKGAAT SVSNPRGRPS RGRPPKLQRN SRGGQGRGVE KPPHLAALIL ARGGSKGIPL KNIKHLAGVP LIGWVLRAAL DSGAFQSVWV STDHDEIENV AKQFGAQVHR RSSEVSKDSS TSLDAIIEFL NYHNEVDIVG NIQATSPCLH PTDLQKVAEM IREEGYDSVF SVVRRHQFRW SEIQKGVREV TEPLNLNPAK RPRRQDWDGE LYENGSFYFA KRHLIEMGYL QGGKMAYYEM RAEHSVDIDV DIDWPIAEQR VLR; Positive Samples: U-87MG, HT-29, 293T, HepG2, DU145, Mouse heart, Rat liver; Cellular Location: Nucleus.

Catalyzes the activation of N-acetylneuraminic acid (NeuNAc) to cytidine 5'-monophosphate N-acetylneuraminic acid (CMP-NeuNAc), a substrate required for the addition of sialic acid. Has some activity toward NeuNAc, N-glycolylneuraminic acid (Neu5Gc) or 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMAS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CMAS
Gene Accession No. P69060, Q8NFW8, Q99KK2
Gene Alias AW208911; Cmas; CMP-N-acetylneuraminic acid synthase; CMP-N-acetylneuraminic acid synthetase; CMPNeu5Ac; CMP-Neu5Ac synthase; CMP-Neu5Ac synthetase; CMP-NeuNAc synthase; CMP-NeuNAc synthetase; CMP-sialic acid synthetase; CSS; cytidine 5'-monophosphate N-acetylneuraminic acid synthetase; cytidine monophosphate N-acetylneuraminic acid synthetase; cytidine monophospho-N-acetylneuraminic acid synthetase; D6Bwg0250e; N-acylneuraminate cytidylyltransferase
Gene Symbols CMAS
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12764, 312826, 55907
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.