Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117876 Supplier Invitrogen™ Supplier No. PA5117876
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CMAS Polyclonal Antibody for Western Blot, IP

Catalyzes the activation of N-acetylneuraminic acid (NeuNAc) to cytidine 5'-monophosphate N-acetylneuraminic acid (CMP-NeuNAc), a substrate required for the addition of sialic acid. Has some activity toward NeuNAc, N-glycolylneuraminic acid (Neu5Gc) or 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMAS
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CMAS
Gene Accession No. Q8NFW8
Gene Alias AW208911; Cmas; CMP-N-acetylneuraminic acid synthase; CMP-N-acetylneuraminic acid synthetase; CMPNeu5Ac; CMP-Neu5Ac synthase; CMP-Neu5Ac synthetase; CMP-NeuNAc synthase; CMP-NeuNAc synthetase; CMP-sialic acid synthetase; CSS; cytidine 5'-monophosphate N-acetylneuraminic acid synthetase; cytidine monophosphate N-acetylneuraminic acid synthetase; cytidine monophospho-N-acetylneuraminic acid synthetase; D6Bwg0250e; N-acylneuraminate cytidylyltransferase
Gene Symbols CMAS
Host Species Rabbit
Immunogen E. coli-derived Human CMAS fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55907
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.