Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CMBL Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299788G
Description
CMBL Polyclonal specifically detects CMBL in Human samples. It is validated for Western Blot, ELISA.Specifications
CMBL | |
Polyclonal | |
Western Blot, ELISA | |
carboxymethylenebutenolidase homolog, carboxymethylenebutenolidase homolog (Pseudomonas), carboxymethylenebutenolidase-like (Pseudomonas), EC 3.1, FLJ23617 | |
Produced in rabbits immunized with purified, recombinant CMBL (Q96DG6; Met1-Met245). | |
0.1 mL | |
metabolism, Proteases & Other Enzymes | |
134147 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction