Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMBL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143929
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143929 Supplier Invitrogen™ Supplier No. PA5143929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human PC-3 whole cell, human HepG2 whole cell, human HEK293 whole cell, human A549 whole cell, human U87 whole cell, rat liver tissue, rat kidney tissue, mouse kidney tissue. IHC: human adnexal serous adenocarcinoma tissue, human lung cancer tissue, human colorectal cancer tissue. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

CMBL is a cysteine hydrolase of the dienelactone hydrolase family that is highly expressed in liver cytosol. CMBL preferentially cleaves cyclic esters, and it activates medoxomil-ester prodrugs in which the medoxomil moiety is linked to an oxygen atom.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMBL
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene Cmbl
Gene Accession No. Q7TP52, Q8R1G2, Q96DG6
Gene Alias 2310016A09Rik; Ab2-225; carboxymethylenebutenolidase homolog; carboxymethylenebutenolidase homolog (Pseudomonas); carboxymethylenebutenolidase-like (Pseudomonas); CMBL; JS-1; Liver regeneration-related protein LRRG072; RGD1306952
Gene Symbols Cmbl
Host Species Rabbit
Immunogen E.coli-derived human LOC134147/CMBL recombinant protein (Position: M1-M246).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134147, 310201, 69574
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.