Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560329
Description
Immunogen sequence: RCSEQVQDFT KCCKNSGVLM VVKCRKENSA LKECLTAYYN DPAFYEECKM EYLKEREEFR KTGIPTKKRL QKLPTSM Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Specifications
CMC1 | |
Polyclonal | |
Unconjugated | |
CMC1 | |
2010110K16Rik; 2010312P05Rik; AI596001; C3orf68; Cmc1; Cmc1p; COX assembly mitochondrial protein 1; COX assembly mitochondrial protein 1 homolog; COX assembly mitochondrial protein homolog; C-x(9)-C motif containing 1; CX9C mitochondrial protein required for full expression of COX 1; C-X9-C motif containing 1; mitochondrial metallochaperone-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z7K0 | |
CMC1 | |
Recombinant protein corresponding to Human CMC1. Recombinant protein control fragment (Product #RP-100278). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction