Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561111 Supplier Invitrogen™ Supplier No. PA561111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPQKDPCQKQ ACEIQKCLQA NSYMESKCQA VIQELRKCCA QYPKGRSVVC SGFEKEEEEN LTRKSASK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 66%.

This gene was identified by involvement in some t(X;14) translocations associated with mature T-cell proliferations. This region has a complex gene structure, with a common promoter and 5' exon spliced to two different sets of 3' exons that encode two different proteins. This gene represents the downstream 8 kDa protein that localizes to mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMC4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMC4
Gene Accession No. P56277
Gene Alias C6.1B; Cmc4; C-x(9)-C motif containing 4 homolog; C-X9-C motif containing 4; Cx9C motif-containing protein 4; mature T-cell proliferation 1 neighbor; mature T-cell proliferation 1 neighbor protein; mature T-cell proliferation 1, isoform p8; mature T-cell proliferation-1 type A; MTCP1; MTCP-1 type A; MTCP1B; MTCP1NB; p8; p8MTCP1; Protein p8 MTCP-1
Gene Symbols CMC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMC4. Recombinant protein control fragment (Product #RP-95191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100272147
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.