Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531131
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (90%), Rat (90%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a core intraflagellar transport (IFT) protein which belongs to a multi-protein complex involved in the transport of ciliary proteins along axonemal microtubules. IFT proteins are found at the base of the cilium as well as inside the cilium, where they assemble into long arrays between the ciliary base and tip. This protein, together with intraflagellar transport protein 81, binds and transports tubulin within cilia and is required for ciliogenesis. Naturally occurring mutations in this gene are associated with amyotrophic lateral sclerosis--frontotemporal dementia and Bardet-Biedl Syndrome.
Specifications
| CMG1 | |
| Polyclonal | |
| Unconjugated | |
| Ift74 | |
| 1700029H06Rik; b2b796Clo; capillary morphogenesis gene 1 protein; capillary morphogenesis protein 1; CCDC2; CMG1; CMG-1; coiled-coil domain containing 2; coiled-coil domain-containing protein 2; Ift74; intraflagellar transport 74; intraflagellar transport 74 homolog; intraflagellar transport protein 74 homolog; LOW QUALITY PROTEIN: intraflagellar transport protein 74 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80173 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.88 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96LB3 | |
| Ift74 | |
| Recombinant protein encompassing a sequence within the center region of human CMG1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction