Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588699 Supplier Invitrogen™ Supplier No. PA588699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASNHKSSAA RPVSRGGVGL TGRPPSGIRP LSGNIRVATA MPPGTARPGS RGCPIGTGGV LSSQIKVAHR PVTQQGLTGM KTGTKGPQRQ ILDKSYYLGL LRSKISELTT EVNKLQKGIE MYNQENSVYL SYEKRAETLA VEIKELQGQL ADYNMLVDKL NTNTEMEEVM NDYNMLKAQN DRETQSLDVI FTERQAKEKQ IRSVEEEIEQ EKQATDDIIK NMSFENQVKY LEMKTTNEKL LQELDTLQQQ LDSQNMKKES LEAEIAHSQV KQEAVLLHEK LYELESHRDQ MIAEDKSIGS PMEEREKLLK QIKDDNQEIA SMERQLTDTK EKINQFIEEI RQLDMDLEEH QDPTNYGWKI LEKNTGSFKK QV; Positive Samples: 293T, A549, Mouse testis; Cellular Location: Cell projection, Cytoplasmic vesicle, cilium.

This gene encodes a core intraflagellar transport (IFT) protein which belongs to a multi-protein complex involved in the transport of ciliary proteins along axonemal microtubules. IFT proteins are found at the base of the cilium as well as inside the cilium, where they assemble into long arrays between the ciliary base and tip. This protein, together with intraflagellar transport protein 81, binds and transports tubulin within cilia and is required for ciliogenesis. Naturally occurring mutations in this gene are associated with amyotrophic lateral sclerosis--frontotemporal dementia and Bardet-Biedl Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ift74
Gene Accession No. Q8BKE9, Q96LB3
Gene Alias 1700029H06Rik; b2b796Clo; capillary morphogenesis gene 1 protein; capillary morphogenesis protein 1; CCDC2; CMG1; CMG-1; coiled-coil domain containing 2; coiled-coil domain-containing protein 2; Ift74; intraflagellar transport 74; intraflagellar transport 74 homolog; intraflagellar transport protein 74 homolog; LOW QUALITY PROTEIN: intraflagellar transport protein 74 homolog
Gene Symbols Ift74
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-372 of human IFT74 (NP_0010926941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67694, 80173
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.