Learn More
Invitrogen™ CMG1 Polyclonal Antibody

Description
Immunogen sequence: KIKKLHQERM ILSTHRNAFK KIMEKQNIEY EALKTQLQEN ETHSQLTNLE RKWQHLEQNN FAMKEFIATK SQESDYQPIK KNVTKQIAEY NKTIVDALHS TSG Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 84%.
Specifications
Specifications
| Antigen | CMG1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Ift74 |
| Gene Accession No. | Q96LB3 |
| Gene Alias | 1700029H06Rik; b2b796Clo; capillary morphogenesis gene 1 protein; capillary morphogenesis protein 1; CCDC2; CMG1; CMG-1; coiled-coil domain containing 2; coiled-coil domain-containing protein 2; Ift74; intraflagellar transport 74; intraflagellar transport 74 homolog; intraflagellar transport protein 74 homolog; LOW QUALITY PROTEIN: intraflagellar transport protein 74 homolog |
| Gene Symbols | Ift74 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.