Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMKLR1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980741
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0741 Supplier Bioss Supplier No. BS10185R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Chemokine-like receptor 1 (CMKLR1) is a receptor for the chemoattractant adipokine chemerin/RARRES2 and for the omega-3 fatty acid derived molecule resolvin E1. Interaction with RARRES2 induces activation of intracellular signaling molecules, such as SKY, MAPK1/3 (ERK1/2), MAPK14/P38MAPK and PI3K leading to multifunctional effects, like, reduction of immune responses, enhancing of adipogenesis and angionesis. Resolvin E1 down-regulates cytokine production in macrophages by reducing the activation of MAPK1/3 (ERK1/2) and NF-kappa-B. CMKLR1 positively regulates adipogenesis and adipocyte metabolism and acts as a coreceptor for several SIV strains (SIVMAC316, SIVMAC239, SIVMACL7E-FR and SIVSM62A), as well as a primary HIV-1 strain (92UG024-2).

Specifications

Antigen CMKLR1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CMKLR1
Gene Accession No. Q99788
Gene Alias chemerin chemokine-like receptor 1; chemerin receptor; CHEMERINR; chemokine receptor-like 1; Chemokine-like receptor 1; CHEMR23; Cmklr1; DEZ; Gpcr27; G-protein coupled chemoattractant-like receptor; G-protein coupled receptor 27; G-protein coupled receptor ChemR23; G-protein coupled receptor DEZ; mcmklr1; orphan G-protein coupled receptor, Dez; resolvin E1 receptor; RVER1
Gene Symbols CMKLR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse CMKLR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1240
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.