Learn More
Invitrogen™ CML66 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113867
Description
Immunogen sequence: EVAANCSLRV KRPLLDPRFE GYKLSLEPLP CYQLELDAAV AEVKLRDDQY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Specifications
CML66 | |
Polyclonal | |
Unconjugated | |
NUDCD1 | |
4921532K09Rik; AA407246; AW260430; AW556235; chronic myelogenous leukemia tumor antigen 66; CML66; CML66-L; NudC domain containing 1; nudC domain-containing protein 1; Nudcd1; OVA66; RGD1310624; Tumor antigen CML66 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84955 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96RS6 | |
NUDCD1 | |
Synthetic peptide directed towards the N terminal region of human NUDCD1 (aa 2-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.