Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CML66 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530857
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CML66, also known as TSGA10 (Testis Specific Gene A10), is a gene located on chromosome 8q23 that encodes a broadly immunogenic tumor antigen. The gene is expressed in various leukemias and solid tumors but has restricted expression in normal tissues, being predominantly found in the testis and heart. The physiological function of CML66 remains unclear. However, its immunogenic properties, including the ability to elicit strong humoral immune responses, make it a potential target for cancer immunotherapy. Studies have identified cytotoxic T lymphocyte (CTL) epitopes derived from CML66, suggesting that TSGA10-targeted therapies could be feasible. Moreover, research into the gene has revealed the existence of different isoforms, such as CML66-L (long) and CML66-S (short), which may have distinct roles in tumorigenesis and immune response modulation.
Specifications
| CML66 | |
| Polyclonal | |
| Unconjugated | |
| NUDCD1 | |
| 4921532K09Rik; AA407246; AW260430; AW556235; chronic myelogenous leukemia tumor antigen 66; CML66; CML66-L; NudC domain containing 1; nudC domain-containing protein 1; Nudcd1; OVA66; RGD1310624; Tumor antigen CML66 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84955 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96RS6 | |
| NUDCD1 | |
| Recombinant protein encompassing a sequence within the center region of human CML66. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction