Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CMP kinase Antibody (08), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306539JF549
Description
CMP kinase Monoclonal antibody specifically detects CMP kinase in Human samples. It is validated for Western BlotSpecifications
CMP kinase | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
08 | |
Western Blot | |
CMKEC 2.7.4.14, CMPKUMKUMP-CMP kinase, cytidine monophosphate (UMP-CMP) kinase 1, cytosolic, Cytidine monophosphate kinase, Cytidylate kinaseUMP/CMP kinase, Deoxycytidylate kinase, EC 2.7.4, UMP/CMPK, UMP-CMPK, UMPKUCK, Uridine monophosphate kinase, Uridine monophosphate/cytidine monophosphate kinase | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CMP kinase (P30085; Met1-Gly196). | |
0.1 mL | |
Proteases & Other Enzymes | |
51727 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction