Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CMP kinase Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300262JF646
Description
CMP kinase Polyclonal antibody specifically detects CMP kinase in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
CMP kinase | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
CMKEC 2.7.4.14, CMPKUMKUMP-CMP kinase, cytidine monophosphate (UMP-CMP) kinase 1, cytosolic, Cytidine monophosphate kinase, Cytidylate kinaseUMP/CMP kinase, Deoxycytidylate kinase, EC 2.7.4, UMP/CMPK, UMP-CMPK, UMPKUCK, Uridine monophosphate kinase, Uridine monophosphate/cytidine monophosphate kinase | |
Produced in rabbits immunized with purified, recombinant Human CMP kinase (Accession#: P30085; Met1-Gly196) | |
0.1 mL | |
Proteases & Other Enzymes | |
51727 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction