Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CMPD-1
GREENER_CHOICE

Catalog No. 218610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-861-0 10 mg
21-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-861-0 Supplier Tocris Bioscience™ Supplier No. 2186/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of p38α-mediated MK2a phosphorylation; also tubulin polymerization inhibitor

CMPD-1 is a non-ATP-competitive, selective inhibitor of p38α-mediated MK2a (mitogen-activated protein kinase-2a) phosphorylation (apparent Ki = 330 nM). Does not inhibit p38α-mediated phosphorylation of MBP and ATF-2. Also tubulin polymerization inhibitor. Cytotoxic in glioblastoma cells at concentrations that do not affect the MK2 pathway.

Specifications

Quantity 10 mg
Product Type CMPD-1
Purity >99%
Molecular Weight (g/mol) 349.4
Inhibitors CMPD-1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.