Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CMPI hydrochloride
GREENER_CHOICESDP

Catalog No. 596310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-631-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-631-0 Supplier Tocris Bioscience™ Supplier No. 5963/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent positive allosteric modulator of α4β2 nAChRs; also inhibitor of (α4)2(β2)3, muscle-type and Torpedo nAChRs

Potent positive allosteric modulator of α4β2 nAChRs (EC50 values are 20 and 18 nM for rat and human, respectively). Selective for hα4β2 over hα3β2, hα3β4 and hα7. Inhibits (α4)2(β2)3, muscle-type and Torpedo nAChRs (IC50 values are 0.5, 0.7 and 0.2 μM, respectively), but not (α4)3(β2)2 receptors. Exhibits ability to photoincorporate into aliphatic and nucleophilic amino acid side chains.

Chemical Identifiers

CAS 2250025-94-4

Specifications

Quantity 10 mg
Solubility Soluble to 50mM in water and to 100mM in DMSO
Formula Weight 379.28
Physical Form Solid
Purity >98%
Chemical Name or Material 3-(2-Chlorophenyl)-5-(5-methyl-1-(piperidin-4-yl)-1H-pyrazol-4-yl)isoxazole hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.