Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMPK1 Monoclonal Antibody (OTI1H1), TrueMAB™, OriGene
SDP

Catalog No. 502539744
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9744 30 μL
50-167-5860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9744 Supplier OriGene Supplier No. TA505375S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CMPK1 Monoclonal antibody specifically detects CMPK1 in Human samples. It is validated for Western Blot

Specifications

Antigen CMPK1
Applications Western Blot
Classification Monoclonal
Clone OTI1H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ETS2
Gene Accession No. P15036
Gene Alias 0610011D08Rik; CK; CK {ECO:0000255; CMK; Cmpk; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidine/uridine monophosphate kinase 1; cytidylate kinase; cytidylate kinase 1; dCMP kinase; dCMP kinase {ECO:0000255; deoxycytidylate kinase; deoxycytidylate kinase {ECO:0000255; EC 2.7.4.14; HAMAP-Rule:MF_03172}; KCY; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); nucleoside-diphosphate kinase; nucleoside-diphosphate kinase {ECO:0000255; RGD1310116; RP11-511I2.1; Uck; Umk; UMP/CMP kinase; UMP/CMP kinase {ECO:0000255; UMP/CMPK; UMP/CMPK {ECO:0000255; ump-cmp; UMP-CMP kinase; UMP-CMP kinase {ECO:0000255; UMP-CMP kinase 1; UMP-CMPk; UMPK; uridine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase {ECO:0000255
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 90-349 of human ETS2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2114
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.