Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMPK1 Monoclonal Antibody (OTI2A2), TrueMAB™, OriGene
SDP

Catalog No. 501675853
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-5853 100 μL
50-253-9748 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-5853 Supplier OriGene Supplier No. TA505366
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Catalyzes the phosphorylation of pyrimidine nucleoside monophosphates at the expense of ATP. Plays an important role in de novo pyrimidine nucleotide biosynthesis. Has preference for UMP and CMP as phosphate acceptors. Also displays broad nucleoside diphosphate kinase activity.

Specifications

Antigen CMPK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CMPK1
Gene Accession No. P30085, Q9DBP5
Gene Alias CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
Gene Symbols CMPK1
Host Species Mouse
Immunogen Full length human recombit protein of human CMPK1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51727, 610291, 66588
Target Species Canine, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.