Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMPK1 Monoclonal Antibody (OTI2F5), TrueMAB™, OriGene
SDP

Catalog No. 502539726
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9726 30 μL
50-167-5887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9726 Supplier OriGene Supplier No. TA505409S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CMPK1 Monoclonal antibody specifically detects CMPK1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CMPK1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ARMC1
Gene Accession No. Q9NVT9
Gene Alias 0610011D08Rik; CK; CK {ECO:0000255; CMK; Cmpk; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidine/uridine monophosphate kinase 1; cytidylate kinase; cytidylate kinase 1; dCMP kinase; dCMP kinase {ECO:0000255; deoxycytidylate kinase; deoxycytidylate kinase {ECO:0000255; EC 2.7.4.14; HAMAP-Rule:MF_03172}; KCY; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); nucleoside-diphosphate kinase; nucleoside-diphosphate kinase {ECO:0000255; RGD1310116; RP11-511I2.1; Uck; Umk; UMP/CMP kinase; UMP/CMP kinase {ECO:0000255; UMP/CMPK; UMP/CMPK {ECO:0000255; ump-cmp; UMP-CMP kinase; UMP-CMP kinase {ECO:0000255; UMP-CMP kinase 1; UMP-CMPk; UMPK; uridine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase {ECO:0000255
Host Species Mouse
Immunogen Full length human recombinant protein of human ARMC1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55156
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.