Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMPK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580488
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CMPK1.
SNX32 (sorting nexin-32), also known as SNX6B (sorting nexin-6B), is a 403 amino acid protein that contains one PX (phox homology) domain and belongs to the sorting nexin family. Existing as two alternatively spliced isoforms, SNX32 may be involved in several stages of intracellular trafficking.
Specifications
CMPK1 | |
Polyclonal | |
Unconjugated | |
Cmpk1 | |
0610011D08Rik; CK; CK {ECO:0000255; CMK; Cmpk; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidine/uridine monophosphate kinase 1; cytidylate kinase; cytidylate kinase 1; dCMP kinase; dCMP kinase {ECO:0000255; deoxycytidylate kinase; deoxycytidylate kinase {ECO:0000255; EC 2.7.4.14; HAMAP-Rule:MF_03172}; KCY; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); nucleoside-diphosphate kinase; nucleoside-diphosphate kinase {ECO:0000255; RGD1310116; RP11-511I2.1; Uck; Umk; UMP/CMP kinase; UMP/CMP kinase {ECO:0000255; UMP/CMPK; UMP/CMPK {ECO:0000255; ump-cmp; UMP-CMP kinase; UMP-CMP kinase {ECO:0000255; UMP-CMP kinase 1; UMP-CMPk; UMPK; uridine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase {ECO:0000255 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P30085 | |
Cmpk1 | |
Recombinant Human CMPK1 protein (Met1-Gly196). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction