Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMPK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584170 Supplier Invitrogen™ Supplier No. PA584170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLQGWNKTMD GKADVSFVLF FDCNNEICIE RCLERGKSSG RSDDNRESLE KRIQTYLQST KPIIDLYEEM GKVKK Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 99%.

SNX32 (sorting nexin-32), also known as SNX6B (sorting nexin-6B), is a 403 amino acid protein that contains one PX (phox homology) domain and belongs to the sorting nexin family. Existing as two alternatively spliced isoforms, SNX32 may be involved in several stages of intracellular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMPK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cmpk1
Gene Accession No. P30085
Gene Alias 0610011D08Rik; CK; CK {ECO:0000255; CMK; Cmpk; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidine/uridine monophosphate kinase 1; cytidylate kinase; cytidylate kinase 1; dCMP kinase; dCMP kinase {ECO:0000255; deoxycytidylate kinase; deoxycytidylate kinase {ECO:0000255; EC 2.7.4.14; HAMAP-Rule:MF_03172}; KCY; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); nucleoside-diphosphate kinase; nucleoside-diphosphate kinase {ECO:0000255; RGD1310116; RP11-511I2.1; Uck; Umk; UMP/CMP kinase; UMP/CMP kinase {ECO:0000255; UMP/CMPK; UMP/CMPK {ECO:0000255; ump-cmp; UMP-CMP kinase; UMP-CMP kinase {ECO:0000255; UMP-CMP kinase 1; UMP-CMPk; UMPK; uridine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase; uridine monophosphate/cytidine monophosphate kinase {ECO:0000255
Gene Symbols Cmpk1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMPK1. Recombinant protein control fragment (Product #RP-94361).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.