Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMPK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534461
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534461 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534461 Supplier Thermo Scientific Supplier No. PA534461
Add to Cart
Edge
Add to Cart

CMPK2 Polyclonal Antibody

CMPK2 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen CMPK2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CMPK2
Gene Accession No. Q3U5Q7, Q5EBM0
Gene Alias 1200004E04Rik; Cmpk2; cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial; cytidine/uridine monophosphate kinase 2; cytidylate kinase 2; mitochondrial UMP-CMP kinase; NDK; Nucleoside-diphosphate kinase; TDKI; thymidine monophosphate kinase 2; thymidylate kinase family LPS-inducible member; thymidylate kinase homologue; thymidylate kinase LPS-inducible member; TMPK2; Tyki; UMP/CMP kinase; UMP-CMP kinase 2, mitochondrial; UMP-CMPK2
Gene Symbols CMPK2
Host Species Rabbit
Immunogen A 19 amino acid synthetic peptide near the carboxy terminus of human CMPK2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129607, 22169, 314004
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.