Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMPK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525130
Description
CMPK2 Polyclonal Antibody for Western Blot, IHC (P), Flow
UMP-CMP kinase 2 (CMPK2) is the first nucleoside monophosphate kinase that has been identified in human mitochondria. It is a component of the salvage pathway for nucleotide synthesis that may participate in terminal differentiation of monocytic cells. There are two distinct domains of CMPK2: the N-terminal domain with unknown function and the C-terminal domain with the kinase function, suggesting that CMPK2 may be a bifunctional protein with other biological functions in addition to its UMP-CMP kinase activity. CMPK2 may participate in dUTP and dCTP synthesis and is responsible for phosphorylation of dCMP and dUMP in mitochondria. CMPK2 is actively involved in macrophage activation and the inflammatory response.
Specifications
| CMPK2 | |
| Polyclonal | |
| Unconjugated | |
| CMPK2 | |
| 1200004E04Rik; Cmpk2; cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial; cytidine/uridine monophosphate kinase 2; cytidylate kinase 2; mitochondrial UMP-CMP kinase; NDK; Nucleoside-diphosphate kinase; TDKI; thymidine monophosphate kinase 2; thymidylate kinase family LPS-inducible member; thymidylate kinase homologue; thymidylate kinase LPS-inducible member; TMPK2; Tyki; UMP/CMP kinase; UMP-CMP kinase 2, mitochondrial; UMP-CMPK2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 129607 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5EBM0 | |
| CMPK2 | |
| KLH conjugated synthetic peptide between 142-170 amino acids from the central region of human CMPK2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction