Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMRF-56 Mouse, anti-Human, R718, Clone: CMRF-56, BD OptiBuild™

Catalog No. BDB751896 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751896 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751896 Supplier BD Biosciences Supplier No. 751896
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CMRF-56 monoclonal antibody specifically binds to a 96 kDa, early activation/differentiation antigen expressed on dendritic cells (DCs). The CMRF-56 antigen is expressed on isolated Langerhans cells, dermal DCs, migratory dermal DCs and monocyte derived DCs (GM-CSF, IL-4, TNF-α). Circulating blood DCs are CMRF-56-, but its expression is induced within 6 hrs of in vitro culture. Tonsil and synovial fluid DCs upregulate CMRF-56 expression following short-term in vitro culture. It is not detected on resting peripheral blood leukocytes, but is on a subpopulation of tonsil B cells and on in vitro cultured CD19+ cells. CMRF-56 was generated using the Hodgkin's-derived L428 cell line as immunogen. This reagent could be useful in flow cytometric or immunomagnetic separation procedures and immunohistochemical staining of acetone-fixed frozen tissue sections.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CMRF-56
Applications Flow Cytometry
Classification Monoclonal
Clone CMRF-56
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.