Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMSS1/C3orf26 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569677
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9677 Supplier Bethyl Laboratories, Inc Supplier No. A305154A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CMSS1 (Cms1 Ribosomal Small Subunit Homolog) is a Protein Coding gene. Diseases associated with CMSS1 includeRefractive Error. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.

Specifications

Antigen CMSS1/C3orf26
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CMSS1
Gene Accession No. Q9BQ75
Gene Alias C3orf26, chromosome 3 open reading frame 26, Cms1 ribosomal small subunit homolog, MGC4308
Gene Symbols CMSS1
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84319
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.