Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CMTM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$377.03 - $666.47
Specifications
| Antigen | CMTM1 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CMTM1 Polyclonal specifically detects CMTM1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CMTM1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| chemokine-like factor super family 1, chemokine-like factor superfamily 1, Chemokine-like factor superfamily member 1, chemokine-like factor-like protein CKLFH1, CKLFH, CKLFH1, CKLFH1a, CKLF-like MARVEL transmembrane domain containing 1, CKLF-like MARVEL transmembrane domain-containing protein 1, CKLFSF1, MGC71870 | |
| CMTM1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 113540 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DPEHAKPESSEAPSGNLKQPETAAALASSGSVD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title