Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMTM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584907
Description
Immunogen sequence: ADAMQLNDKW QGLCWPMMDF LR.
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants containing different 5' UTRs, but encoding the same protein, have been identified.
Specifications
CMTM3 | |
Polyclonal | |
Unconjugated | |
CMTM3 | |
9430096L06Rik; AI413895; BNAS2; chemokine-like factor super family 3; chemokine-like factor superfamily member 3; CKLF like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain-containing protein 3; CKLFSF3; CMTM3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MX0 | |
CMTM3 | |
Recombinant protein corresponding to Human CMTM3. Recombinant protein control fragment (Product #RP-108605). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction