Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101524 Supplier Invitrogen™ Supplier No. PA5101524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CKLF3.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants containing different 5' UTRs, but encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CMTM3
Gene Accession No. Q96MX0
Gene Alias 9430096L06Rik; AI413895; BNAS2; chemokine-like factor super family 3; chemokine-like factor superfamily member 3; CKLF like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain-containing protein 3; CKLFSF3; CMTM3
Gene Symbols CMTM3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CMTM3(Accession Q96MX0), corresponding to amino acid residues M1-L31.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123920
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.