Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554914 Supplier Invitrogen™ Supplier No. PA554914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEELDGFEGE ASSTSMISGA SSPYQPTTEP VSQRRGLAGL RCDPDYL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTM4
Gene Accession No. Q8IZR5
Gene Alias chemokine-like factor super family 4; chemokine-like factor superfamily member 4; CKLF like MARVEL transmembrane domain containing 4; CKLF-like MARVEL transmembrane domain containing 4; CKLF-like MARVEL transmembrane domain-containing protein 4; Cklfsf4; Cmtm4; D19397; ENSMUSG00000051554; Gm9853
Gene Symbols CMTM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMTM4. Recombinant protein control fragment (Product #RP-91191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.