Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562589 Supplier Invitrogen™ Supplier No. PA562589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLSARDRRDR HPEEGVVAEL QGFAVDKAFL TSHKGI Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 64%.

This gene belongs to the chemokine-like factor gene superfamily. This family of genes encodes multi-pass membrane proteins that are similar to both the chemokine and the transmembrane 4 superfamilies of signaling molecules. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. Highly expressed in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTM5
Gene Accession No. Q96DZ9
Gene Alias 1500005P16Rik; 2900052H21Rik; chemokine-like factor super family 5; chemokine-like factor super family five variant 4; chemokine-like factor superfamily 5; chemokine-like factor superfamily member 5; CKLF like MARVEL transmembrane domain containing 5; CKLF-like MARVEL transmembrane domain containing 5; CKLF-like MARVEL transmembrane domain-containing protein 5; CKLFSF5; CMTM5
Gene Symbols CMTM5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMTM5. Recombinant protein control fragment (Product #RP-99878).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.