Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMTM5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988208
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8208 Supplier Bioss Supplier No. BS8023R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to the chemokine-like factor gene superfamily. This family of genes encodes multi-pass membrane proteins that are similar to both the chemokine and the transmembrane 4 superfamilies of signaling molecules. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. Highly expressed in the brain.

Specifications

Antigen CMTM5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CMTM5
Gene Accession No. Q96DZ9
Gene Alias 1500005P16Rik; 2900052H21Rik; chemokine-like factor super family 5; chemokine-like factor super family five variant 4; chemokine-like factor superfamily 5; Chemokine-like factor superfamily member 5; CKLF like MARVEL transmembrane domain containing 5; CKLF-like MARVEL transmembrane domain containing 5; CKLF-like MARVEL transmembrane domain-containing protein 5; Cklfsf5; Cmtm5
Gene Symbols CMTM5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CKLFSF5/CMTM5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116173
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.