Learn More
Invitrogen™ CMTM7 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CMTM7 protein.
Specifications
Specifications
| Antigen | CMTM7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CMTM7 |
| Gene Accession No. | Q96FZ5 |
| Gene Alias | AI481279; chemokine-like factor super family 7; chemokine-like factor super family member 7 variant 2; chemokine-like factor superfamily 7; chemokine-like factor superfamily member 7; CKLF like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain-containing protein 7; Cklfsf7; Cmtm7; LNV |
| Gene Symbols | CMTM7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.