Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103744 Supplier Invitrogen™ Supplier No. PA5103744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CKLF7.

CMTM7 belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. The protein encoded by this gene is highly expressed in leukocytes, but its exact function is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CMTM7
Gene Accession No. Q96FZ5, Q9ESD6
Gene Alias AI481279; chemokine-like factor super family 7; chemokine-like factor super family member 7 variant 2; chemokine-like factor superfamily 7; chemokine-like factor superfamily member 7; CKLF like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain-containing protein 7; Cklfsf7; Cmtm7; LNV
Gene Symbols CMTM7
Host Species Rabbit
Immunogen A synthesized peptide derived from human CMTM7(Accession Q96FZ5), corresponding to amino acid residues S111-S161.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102545, 112616
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.