Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMTM8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550502
Description
The antibody detects endogenous levels of total CMTM8 protein.
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown.
Specifications
CMTM8 | |
Polyclonal | |
Unconjugated | |
CMTM8 | |
2700018N07Rik; AA408515; chemokine-like factor super family 8; chemokine-like factor superfamily 8; chemokine-like factor superfamily member 8; chemokine-like related protein 8; CKLF like MARVEL transmembrane domain containing 8; CKLF-like MARVEL transmembrane domain containing 8; CKLF-like MARVEL transmembrane domain-containing protein 8; CKLFSF8; CKLFSF8-V2; Cmtm8; Unknown (protein for MGC:140079) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152189 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IZV2 | |
CMTM8 | |
Synthetic peptide corresponding to residues near the N terminal of human CKLF-like MARVEL transmembrane domain containing 8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction