Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563061 Supplier Invitrogen™ Supplier No. PA563061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARSHTVTTTA SSFAENFSTS SSSFAYDREF LRTL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTM8
Gene Accession No. Q8IZV2
Gene Alias 2700018N07Rik; AA408515; chemokine-like factor super family 8; chemokine-like factor superfamily 8; chemokine-like factor superfamily member 8; chemokine-like related protein 8; CKLF like MARVEL transmembrane domain containing 8; CKLF-like MARVEL transmembrane domain containing 8; CKLF-like MARVEL transmembrane domain-containing protein 8; CKLFSF8; CKLFSF8-V2; Cmtm8; Unknown (protein for MGC:140079)
Gene Symbols CMTM8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMTM8. Recombinant protein control fragment (Product #RP-99893).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.