Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMYA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140296
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FISEVSREDY GKKEISGDSE EMNINSVVTS ADGENLEIQS YSLIGEKLVM EEAKTIVPPH VTDSKRVQKP AIAPPSK.
SPRYD2, also known as Myospryn, was originally identified as the muscle-specific partner of dysbindin and as a Mef-2 target gene. It is a large scaffolding protein localized to the Z-disc/costamere region of striated muscle. SPRYD2 includes a noncanonical tripartite motif (TRIM-like) that lacks the RING domain but consists of a B-box coiled coil (BBC), fibronectin 3 (FN3) repeats, and SPRY domains. SPRYD2 interacts with desmin and calcineurin, and it has been suggested to play a role in the biogenesis of lysosome and negatively regulates slow-fiber-type transformation and skeletal muscle regeneration. SPRYD2 is dysregulated in Duchenne muscular dystrophy.
Specifications
CMYA5 | |
Polyclonal | |
Unconjugated | |
CMYA5 | |
2310076E16Rik; 2310076E21Rik; AA420382; AI504003; C5orf10; cardiomyopathy associated 5; cardiomyopathy-associated protein 5; CMYA5; DTNBP2; Dystrobrevin-binding protein 2; genethonin-3; heart and skeletal muscle-specific and sprouty domain-containing; myospryn; SPRY domain-containing protein 2; SPRYD2; sr553; Srfsd; stretch response protein 553; stretch-response protein 553; stretch-responsive fibronectin protein type 3; Tims; TRIM76; TRIM-like protein; tripartite motif-containing 76; tripartite motif-containing protein 76 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3K9 | |
CMYA5 | |
Recombinant protein corresponding to Human CMYA5. Recombinant protein control fragment (Product #RP-109651). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction