Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CMYA5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317135100UL
This item is not returnable.
View return policy
Description
CMYA5 Polyclonal antibody specifically detects CMYA5 in Human samples. It is validated for ImmunofluorescenceSpecifications
CMYA5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
C5orf10, cardiomyopathy associated 5, cardiomyopathy-associated protein 5, DKFZp451G182, DKFZp451G223, DTNBP2, dystrobrevin-binding protein 2, genethonin-3, myospryn, SPRY domain-containing protein 2, SPRYD2, tripartite motif-containing protein 76 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: SDVPKQSVLVSKHHLEAAEDTRVKEPLSSAKSNYAQFISNTSASNADKMVSNKEMPKEPEDTYAKGEDFTVTSKPAGLS | |
100 μg | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
202333 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction