Learn More
Thermo Scientific™ CN Selective Supplement (Cefsulodin, Novobiocin)
Catalog No. p-6092039
Click to view available options
Form:
Liquid
Quantity:
1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Selectively isolate Yersinia enterocolitica using CN Selective Supplement (Cefsulodin, Novobiocin).
Selectively isolate Yersinia enterocolitica using Thermo Scientific™ Remel™ CN Selective Supplement (Cefsulodin, Novobiocin). This supplement is used in the preparation of CIN agar base which makes it selective and a differential medium for isolation of Y. enterocolitica. Supplementation with CN supplement improves inhibition of normal enteric organisms such as Escherichia coli, Klebsiella pneumoniae, Proteus mirabilis and Pseudomonas aeruginosa, as well as gram-positive organisms.
Selectively isolate Yersinia enterocolitica using CN Selective Supplement (Cefsulodin, Novobiocin).
The complete medium of CIN Agar formulation of Schiemann contains this highly selective supplement which is recommended for use in the isolation and cultivation of Yersinia enterocolitica from clinical specimens and food.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type Selective Supplement
Quantity 1 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.