Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CNAP1 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CNAP1 Polyclonal specifically detects CNAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
CNAP1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
Human | |
CAPD2, CAP-D2, Chromosome condensation-related SMC-associated protein 1, Chromosome-associated protein D2, CNAP1XCAP-D2 homolog, hCAP-D2condensin complex subunit 1, KIAA0159chromosome condensation related SMC associated protein 1, Non-SMC condensin I complex subunit D2, non-SMC condensin I complex, subunit D2 | |
NCAPD2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9918 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DKSVLVCKNAIQLLASFLANNPFSCKLSDADLAGPLQKETQKLQEMRAQRRTAAASAVLDPEEEWEAMLPELKSTLQQLLQLPQGEEEIPEQIA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title