Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNAP1 Antibody (SR1189), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322503100UL
Description
CNAP1 Monoclonal antibody specifically detects CNAP1 in Human samples. It is validated for Western BlotSpecifications
CNAP1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1189 | |
Western Blot 1:500-1:2000 | |
CAPD2, CAP-D2, Chromosome condensation-related SMC-associated protein 1, Chromosome-associated protein D2, CNAP1XCAP-D2 homolog, hCAP-D2condensin complex subunit 1, KIAA0159chromosome condensation related SMC associated protein 1, Non-SMC condensin I complex subunit D2, non-SMC condensin I complex, subunit D2 | |
A synthesized peptide derived from human CNAP1 (Uniprot #: Q15021) | |
100 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
9918 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction