Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNBP Antibody (38F10), Alexa Fluor™ 647, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242632AF647

Catalog No. NBP242632AH


Only null left
Add to Cart

Description

Description

CNBP Monoclonal specifically detects CNBP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CNBP
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CNBP
Recombinant human CNBP (1-170aa) purified from E. coli
100 μL
DNA replication Transcription Translation and Splicing
7555
Store at 4C in the dark.
IgG2b κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
38F10
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
CCHC-type zinc finger, nucleic acid binding protein, cellular nucleic acid binding protein, cellular nucleic acid-binding protein, DM2, erythroid differentiation-related, FLJ11631, PROMM, RNF163CNBP1ZNF9ZCCHC22, sterol regulatory element-binding protein, zinc finger protein 273, Zinc finger protein 9, zinc finger protein 9 (a cellular retroviral nucleic acid binding protein)
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.